missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF192 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551942
This item is not returnable.
View return policy
Description
Immunogen sequence: VKEHQLENGE EVVTLLEDLE RQIDILGRPV SARVHGHRVL WEEVVHSASA PEPPNTQLQS EATQHKSPVP QESQERAMST SQSPTRSQKG SSGDQEMTAT LLTAGFQTLE KIED Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 70%.
May be involved in transcriptional regulation. [UniProt].
Specifications
| ZNF192 | |
| Polyclonal | |
| Unconjugated | |
| ZKSCAN8 | |
| 2510038J07Rik; CAML; D430019P06Rik; LD5-1; Zfp192; zinc finger protein 192; Zinc finger protein with KRAB and SCAN domains 8; zinc finger with KRAB and SCAN domains 8; ZKSCAN8; ZNF192; ZSCAN40 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7745 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15776 | |
| ZKSCAN8 | |
| Recombinant protein corresponding to Human ZNF192. Recombinant protein control fragment (Product #RP-99745). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction