missing translation for 'onlineSavingsMsg'
Learn More

ZNF187 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18366016 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Conditionnement:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
18366016 100 μL 100µL
18373206 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18366016 Fournisseur Novus Biologicals Code fournisseur NBP303215100UL

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

ZNF187 Polyclonal specifically detects ZNF187 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation Sequencing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF187
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias MGC2815, Protein SRE-ZBP, Zinc finger and SCAN domain-containing protein 26, zinc finger protein 187, ZSCAN26SRE-ZBP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 105-344 of human ZNF187 (NP_689949.3).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7741
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.