missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF182 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568603
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Zinc-finger proteins bind nucleic acids and play important roles in various cellular functions, including cell proliferation, differentiation, and apoptosis. This gene encodes a zinc finger protein, and belongs to the krueppel C2H2-type zinc-finger protein family. It may be involved in transcriptional regulation. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| ZNF182 | |
| Polyclonal | |
| Unconjugated | |
| ZNF182 | |
| 6330583I20Rik; HHZ150; KOX14; MGC137263 protein; Zfp182; Zinc finger protein 182; zinc finger protein 21; zinc finger protein 21 (KOX 14); zinc finger protein KOX14; ZNF182; ZNF21 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7569 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P17025 | |
| ZNF182 | |
| synthetic peptide directed towards the middle region of human ZNF182. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction