missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF180 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556026
This item is not returnable.
View return policy
Description
Immunogen sequence: QEILLQEVAF TQRKAVIHER VCKSDETGEK SGLNSSLFSS PVIPIRNHFH KHVSHAKKWH LNAAVNSHQK INENETLYEN NECGKPPQSI HLIQFTRTQT KDKCYGFSDR IQSFCHGTPL HIHEKIHGGG KTFDFKECGQ Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 43%.
Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See MIM 604749 for additional information on zinc finger proteins.
Specifications
| ZNF180 | |
| Polyclonal | |
| Unconjugated | |
| ZNF180 | |
| HHZ168; zinc finger protein 180; ZNF180 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UJW8 | |
| ZNF180 | |
| Recombinant protein corresponding to Human ZNF180. Recombinant protein control fragment (Product #RP-99725). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction