missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552259
This item is not returnable.
View return policy
Description
Immunogen sequence: ISIQVLGQDI LSEKMESPSC QVGEVEPHLE VVPQELGLEN SSSGPGELLS HIVKEESDTE AELALAASQP ARLEERLIRD QDLGASLLPA APQEQWRQLD STQKEQYWDL MLETYGKMVS GAGISHPKSD LTNS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 69%.
ZNF18 be involved in transcriptional regulation.
Specifications
| ZNF18 | |
| Polyclonal | |
| Unconjugated | |
| ZNF18 | |
| 1700128E15Rik; D11Ertd714e; HDSG1; Heart development-specific gene 1 protein; KOX11; Zfp18; Zfp535; Zinc finger protein 18; zinc finger protein 18 (KOX 11); zinc finger protein 535; Zinc finger protein KOX11; Zinc finger protein with KRAB and SCAN domains 6; zinc finger with KRAB and SCAN domains 6; Zkscan6; Znf18; ZNF535; ZSCAN38 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7566 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17022 | |
| ZNF18 | |
| Recombinant protein corresponding to Human ZNF18. Recombinant protein control fragment (Product #RP-89371). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction