missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF179 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA553402

Product Code. 15761341

  • £481.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LAQEIKNLSG WMGRTGPGFT SPDEMAAQLH DLRKVEAAKR EFEEYVRQQD VATKRIFSAL RVLPDTMRNL LSTQKDAILA RHGVALLCKG RDQTLEALEA ELQATAKAFM DSYT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

The specific function of this protein remains unknown. This gene encodes a member of the RING finger protein family of transcription factors. The protein is primarily expressed in brain. The gene is located within the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF179
Polyclonal
Unconjugated
RNF112
AI854096; Bfb; bfp; Brain finger protein; LOW QUALITY PROTEIN: RING finger protein 112; neurolastin; R75574; RING finger protein 112; Rnf112; Zfp179; zinc finger protein 179; ZNF179
Rabbit
Antigen affinity chromatography
RUO
7732
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9ULX5
RNF112
Recombinant protein corresponding to Human ZNF179. Recombinant protein control fragment (Product #RP-90951).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.