missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF179 Polyclonal Antibody
Description
Peptide sequence: REFEEYVRQQ DVATKRIFSA LRVLPDTMRN LLSTQKDAIL ARHGVALLCK Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.
The specific function of this protein remains unknown. This gene encodes a member of the RING finger protein family of transcription factors. The protein is primarily expressed in brain. The gene is located within the Smith-Magenis syndrome region on chromosome 17.
Specifications
Specifications
| Antigen | ZNF179 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | RNF112 |
| Gene Accession No. | Q9ULX5 |
| Gene Alias | AI854096; Bfb; bfp; Brain finger protein; LOW QUALITY PROTEIN: RING finger protein 112; neurolastin; R75574; RING finger protein 112; Rnf112; Zfp179; zinc finger protein 179; ZNF179 |
| Gene Symbols | RNF112 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?