missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF177 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ZNF177 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF177 Polyclonal specifically detects ZNF177 in Human samples. It is validated for Western Blot.Specifications
| ZNF177 | |
| Polyclonal | |
| Rabbit | |
| NP_003442 | |
| 7730 | |
| Synthetic peptide directed towards the middle region of human ZNF177. Peptide sequence AFRNSSCLRVHVRTHTGEKPYKCFQCEKAFSTSTNLIMHKRIHNGQKLHE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PIGX, zinc finger protein 177 | |
| ZNF177 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title