missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF174, Mouse anti-Human, Clone: 2D7-E9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007727-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MAAKMEITLSSNTEASSKQERHIIAKLEEKRGPPLQKNCPDPELCRQSFRRFCYQEVSGPQEALSQLRQLCRQWLQPELHTKEQILELLVMEQFLTILPPEIQARVRHRCPMSSKEIVTLVEDFHRASKKPKQWVAVCMQGQKVLLEKTGSQLGEQELPDFQPQTPRRDLRESSPAEPSQAGAYDRLSPHHWEKSPLLQEPTPKLAGTELLIEKTDPNMATDELPCKLWLSFIASpecifications
| ZNF174 | |
| Monoclonal | |
| Unconjugated | |
| zinc finger protein 174 | |
| ZSCAN8 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, KnockDown, Western Blot | |
| 2D7-E9 | |
| In 1x PBS, pH 7.4 | |
| BC000876 | |
| ZNF174 | |
| ZNF174 (AAH00876, 1 a.a. ∼ 234 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7727 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction