missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF174 Polyclonal specifically detects ZNF174 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ZNF174 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | AW-1, zinc finger protein 174, ZSCAN8Zinc finger and SCAN domain-containing protein 8 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ZNF174 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?