missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF167 Monoclonal Antibody (3A12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522223
This item is not returnable.
View return policy
Description
Peptide Sequence: TTKESPPTSP LSGGSAPGAH LEPPYDPGTH HLPSGDFAQC TSPVPTLPQV GNSGDQAGAT VLRMVRPQDT VAYEDLSVDY TQKKWKSLTL SQRALQWNMM.
ZNF167, a gene located on chromosome 3, encodes for a zinc finger protein.
Specifications
| ZNF167 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9P0L1 | |
| ZKSCAN7 | |
| ZKSCAN7 (NP_079445, 161 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3A12 | |
| Unconjugated | |
| ZKSCAN7 | |
| ZFP; Zfp167; Zinc finger protein 167; zinc finger protein 448; Zinc finger protein 64; zinc finger protein with KRAB and SCAN domains 7; zinc finger with KRAB and SCAN domains 7; ZKSCAN7; ZNF167; ZNF448; ZNF64; ZSCAN39 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55888 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction