missing translation for 'onlineSavingsMsg'
Learn More

ZNF167 Antibody (3A12), Novus Biologicals™

Product Code. 18410050 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410050 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410050 Supplier Novus Biologicals Supplier No. H00055888M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF167 Monoclonal antibody specifically detects ZNF167 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF167
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3A12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_079445
Gene Alias FLJ12738, ZFP, zinc finger protein 167, Zinc finger protein 448Zinc finger protein with KRAB and SCAN domains 7, Zinc finger protein 64ZNF448, ZKSCAN7ZNF64
Host Species Mouse
Immunogen ZNF167 (NP_079445, 161 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TTKESPPTSPLSGGSAPGAHLEPPYDPGTHHLPSGDFAQCTSPVPTLPQVGNSGDQAGATVLRMVRPQDTVAYEDLSVDYTQKKWKSLTLSQRALQWNMM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55888
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.