missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF165 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48507-25ul
This item is not returnable.
View return policy
Description
ZNF165 Polyclonal antibody specifically detects ZNF165 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF165 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Cancer/testis antigen 53, CT53Zinc finger and SCAN domain-containing protein 7, zinc finger protein 165, ZSCAN7LD65ZPF165 | |
| This antibody was developed against a recombinant protein corresponding to amino acids:AWVHEHYPESGEEAVTILEDLERGTDEAVLQVQAHEHGQEIFQKKVSPPGPALNVKLQPVETKAHFDSSEPQLLWDCDNESENSRSMPKLEIFEKIESQRIISGRISGYISEASGESQ | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7718 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction