missing translation for 'onlineSavingsMsg'
Learn More

ZNF148 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18415543 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415543 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415543 Supplier Novus Biologicals Supplier No. NBP171872UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF148 Polyclonal antibody specifically detects ZNF148 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF148
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BERF-1, BFCOL1, CACCC box-binding protein, CLL-associated antigen KW-10, HT-BETA, pHZ-52, Transcription factor ZBP-89, ZBP89, ZBP-89, ZFP148zinc finger protein 148 (pHZ-52), Zinc finger DNA-binding protein 89, zinc finger protein 148
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 744 and 794 of human Zinc Finger DNA-Binding Protein 89 using the numbering given in entry NP_068799.2 (GeneID 7707).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7707
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.