missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF146 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20385-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF146 (zinc finger protein 146), also known as OZF (only zinc finger protein), is a 292 amino acid nuclear protein that binds DNA and belongs to the Kruppel C2H2-type zinc-finger protein family. ZNF146 interacts with UBC9 (ubiquitin carrier protein 9) and RAP1 (repressor/activator protein 1 homolog), and is highly expressed in multiple pancreatic and colorectal cancers. Lower levels of expression have been observed in kidney, lung, placenta and brain.Specifications
| ZNF146 | |
| Polyclonal | |
| Unconjugated | |
| ZNF146 | |
| Only zinc finger protein, OZF, zinc finger protein 146, Zinc finger protein OZF, ZNF146 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102643033, 103691098, 7705 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15072, Q8BQN6 | |
| ZNF146 | |
| ZNF146 Fusion Protein Ag14223 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction