missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF143 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16618-1-AP-150UL
This item is not returnable.
View return policy
Description
Transcriptional activator. Activates the gene for selenocysteine tRNA (tRNAsec). Binds to the SPH motif of small nuclear RNA (snRNA) gene promoters. Participates in efficient U6 RNA polymerase III transcription via its interaction with CHD8.Specifications
| ZNF143 | |
| Polyclonal | |
| Unconjugated | |
| ZNF143 | |
| hStaf, pHZ 1, SBF, SPH binding factor, STAF, zinc finger protein 143, ZNF143 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20841, 361627, 7702 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70230, P52747, Q5XIU2 | |
| Znf143 | |
| ZNF143 Fusion Protein Ag9914 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction