missing translation for 'onlineSavingsMsg'
Learn More

ZNF143 Antibody (2B4), Novus Biologicals™

Product Code. 18410740 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410740 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410740 Supplier Novus Biologicals Supplier No. H00007702M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF143 Monoclonal antibody specifically detects ZNF143 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF143
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), ChIP Assay
Classification Monoclonal
Clone 2B4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH20219
Gene Alias hStaf, SBFzinc finger protein 143 (clone pHZ-1), Selenocysteine tRNA gene transcription-activating factor, SPH-binding factor, STAFpHZ-1, transcriptional activator Staf, zinc finger protein 143
Host Species Mouse
Immunogen ZNF143 (AAH20219, 1 a.a. ~ 626 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTEFPGGGMEAQHVTLCLTEAVTVADGDNLENMEGVSLQAVTLADGSTAYIQHNSKDAKLIDGQVIQLEDGSAAYVQHVPIPKSTGDSLRLEDGQAVQLEDGTTAFIHHTSKDSYDQSALQAVQLEDGTTAYIHHAVQVPQSDTILAIQADGTVAGLHTGDATIDPDTISALEQYAAKVSIDGSESVAGTGMIGENEQEKKMQIVLQGHATRVTAKSQQSGEKAFRCEYDGCGKLYTTAHHLKVHERSHTGDRPYQCEHAGCGKAFATGYGLKSHVRTHTGEKPYRCSEDNCTKSFKTSGDLQKHIRTHTGERPFKCPFEGCGRSFTTSNIRKVHVRTHTGERPYYCTEPGCGRAFASATNYKNHVRIHTGEKPYVCTVPGCDKRFTEYSSLYKHHVVHTHSKPYNCNHCGKTYKQISTLVMHKRTAHNDTEPIEEEQEAFFEPPPGQGEDVLKGSQITYVTGVEGDDVVSTQVATVTQSGLSQQVTLISQDGTQHVNISQADMQAIGNTITMVTQDGTPITVPAHDAVISSAGTHSVAMVTAEGTEGQQVAIVAQDLAAFHTASSEMGHQQHSHHLVTTETRPLTLVATSNGTQIAVQLGEQPSLEEAIRIASRIQQGETPGLDD
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7702
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.