missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF14 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15709871 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15709871 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15709871 Supplier Invitrogen™ Supplier No. PA562068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: CESRRGSKCG ETTSQMPNVN INKETFTGA Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 44%.

ZNF14 contains a zinc finger and a Kruppel-associated box (KRAB) domain. KRAB domain is known to be involved in the transcriptional repression of a number of zinc finger proteins.The protein encoded by this gene contains a zinc finger and a Kruppel-associated box (KRAB) domain. KRAB domain is known to be involved in the transcriptional repression of a number of zinc finger proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF14
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF14
Gene Accession No. P17017
Gene Alias GIOT4; GIOT-4; gonadotropin inducible transcription repressor-4; gonadotropin-inducible ovary transcription repressor 4; KOX6; Zinc finger protein 14; zinc finger protein 14 (KOX 6); zinc finger protein KOX6; ZNF14
Gene Symbols ZNF14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF14. Recombinant protein control fragment (Product #RP-101299).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7561
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.