missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF135 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552368
This item is not returnable.
View return policy
Description
Immunogen sequence: FLWDGLWYCR GEDTEGHWEW SCESLESLAV PVAFTPVKTP VLEQWQRNGF GENISLNPDL PHQPMTPERQ SPHTWGTRGK REKPDLNVLQ KTCVKEKPYK CQECGKAFSH SSALI Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 31%.
ZNF135 may be involved in transcriptional regulation.
Specifications
| ZNF135 | |
| Polyclonal | |
| Unconjugated | |
| ZNF135 | |
| pHZ-17; pT3; Zinc finger protein 135; zinc finger protein 135 (clone pHZ-17); zinc finger protein 61; Zinc finger protein 78-like 1; zinc finger protein 78-like 1 (pT3); ZNF135; ZNF61; ZNF78L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7694 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P52742 | |
| ZNF135 | |
| Recombinant protein corresponding to Human ZNF135. Recombinant protein control fragment (Product #RP-89430). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction