missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF134 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15770591 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15770591 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15770591 Supplier Invitrogen™ Supplier No. PA552643

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DCGKVFRHKS TLVQHESIHT GENPYDCSDC GKSFGHKYTL IKHQRIHTES KPFECIECGK FFSRSSDYIA HQRVHTGERP FVCSKCGKDF IRTSHLVRHQ RVHTGERPYE CSECGKAYSL SSHLNRHQK Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 63%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc-finger protein 134 (ZNF134) is a 348 amino acid member of the Krueppel C2H2-type zinc-finger protein family. ZNF134 localizes to the nucleus and contains nine C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF134
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF134
Gene Accession No. P52741
Gene Alias pHZ-15; zinc finger protein 134; zinc finger protein 134 (clone pHZ-15); ZNF134
Gene Symbols ZNF134
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF134. Recombinant protein control fragment (Product #RP-89390).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7693
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.