missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF133 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35593-20ul
This item is not returnable.
View return policy
Description
ZNF133 Polyclonal antibody specifically detects ZNF133 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ZNF133 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.1.1, EC 2.1.1.43, pHZ-13, pHZ-66, zinc finger protein 133, zinc finger protein 133 (clone pHZ-13), Zinc finger protein 150, zinc finger protein 150 (pHZ-66), ZNF150 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ZNF133 (NP_003425.2).,, Sequence:, MAFRDVAVDFTQDEWRLLSPAQRTLYREVMLENYSNLVSLGISFSKPELITQLEQGKETWREEKKCSPATCPDPEPELYLDPFCPPGFSSQKFPMQHVLCNHPPWIFTCLCAEGNIQPGDPGPGDQEKQQQASEGRPWSDQAEGPEGEGAMPLFGRTKKRTLGAFSRPPQRQPVSSRNGLRGVELEASPAQTGNPEETDKLLKRIEVLGFGTVNCGECGL | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7692 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction