missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF131 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17118873
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17118873 100 μL 100µL
17199633 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17118873 Supplier Invitrogen™ Supplier No. PA5110947

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GRKKLYECQV CNSVFNSWDQ FKDHLVIHTG DKPNHCTLCD LWFMQGNELR RHLSDAHNIS ERLVTEEVLS VETRVQTEPV TSMTIIEQVG KVHVLPLLQV QVDSAQVTVE QVHPDLLQD Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

May be involved in transcriptional regulation. Plays a role during development and organogenesis as well as in the function of the adult central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF131
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF131
Gene Accession No. P52739
Gene Alias 2610109I01Rik; AW558871; pHZ-10; ZBTB35; Zfp131; zinc finger and BTB domain containing 35; zinc finger protein 131; zinc finger protein 131 (clone pHZ-10); Zinc finger protein 131-like protein; ZNF131
Gene Symbols ZNF131
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ZNF131. Recombinant protein control fragment (Product #RP-105779).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7690
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.