missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF124 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20961
This item is not returnable.
View return policy
Description
ZNF124 Polyclonal specifically detects ZNF124 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZNF124 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| HZF16, HZF-16, MGC117046, zinc finger protein 124, zinc finger protein 124 (HZF-16), Zinc finger protein HZF-16 | |
| Rabbit | |
| 40 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q15973 | |
| ZNF124 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF124. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 7678 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction