Learn More
Invitrogen™ ZNF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589436
Description
Immunogen sequence: EGEFLLQSYP DEVWQTDDLI ERIQEEENKP SRQTVFIETL IEERGNVPGK TFDVETNPVP SRKIAYKNSL CDSCEKCLTS VSEYISSDGS YARMKADECS GCGKSLLHIK LEKTHPGDQA YEFNQNGEPY TLNEESLYQK IRILEKPFEY IECQKAFQKD TVFVNHMEEK P; Positive Samples: Mouse kidney; Cellular Location: Nucleus.
Two members of the human zinc finger Kruppel family, ZNF 12 (KOX 3) and ZNF 26 (KOX 20), have been localized by somatic cell hybrid analysis and in situ chromosomal hybridization. The presence of individual human zinc finger genes in mouse-human hybrid DNAs was correlated with the presence of specific human chromosomes or regions of chromosomes in the corresponding cell hybrids. Analysis of such mouse-human hybrid DNAs assigned the ZNF 12 (KOX 3) gene to chromosome region 7p.
Specifications
| ZNF12 | |
| Polyclonal | |
| Unconjugated | |
| Zfp12 | |
| C530015C18; GIOT3; GIOT-3; gonadotropin inducible transcription repressor 3; gonadotropin-inducible ovary transcription repressor 3; HZF11; KOX3; Krox-7; Zfp12; Zfp-12; zinc finger protein 11; zinc finger protein 12; Zinc finger protein 325; zinc finger protein KOX3; Znf12; ZNF325 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 231866 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TSI0 | |
| Zfp12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-240 of human ZNF12 (NP_0573492). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.