Learn More
ZNF10 Antibody (CL9744) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88939PCP
Product Code. 30302111
  • £558.00 / 0.10mL
Description
ZNF10 Monoclonal antibody specifically detects ZNF10 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF10
Monoclonal
PerCP
KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1
This antibody was developed using a synthetic peptide derived from P21506, with the exact immunogen sequence remaining proprietary.
0.1 mL
Transcription Factors and Regulators
7556
Store at 4⪚C in the dark.
IgG1
Western Blot, Immunocytochemistry/Immunofluorescence
CL9744
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified