missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF10 Antibody (CL9744) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44232
This item is not returnable.
View return policy
Description
ZNF10 Monoclonal antibody specifically detects ZNF10 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF10 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1 | |
| This antibody was generated using a synthetic peptide of P21506, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Transcription Factors and Regulators | |
| 7556 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL9744 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction