missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF10 Monoclonal antibody specifically detects ZNF10 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZNF10 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 980809 |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1 |
| Host Species | Mouse |
| Immunogen | Synthetic peptide containing human ZNF10, Accession # P21506 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?