missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNDR1 Antibody (6C12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00030834-M01
This item is not returnable.
View return policy
Description
ZNDR1 Monoclonal antibody specifically detects ZNDR1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| ZNDR1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DNA-directed RNA polymerase I subunit RPA12, HTEX-6, hZR14, MGC13376, RNA polymerase I small specific subunit Rpa12, Rpa12, tctex-6, TEX6, transcription-associated zinc ribbon protein, zinc ribbon domain containing 1, zinc ribbon domain containing, 1, Zinc ribbon domain-containing protein 1 | |
| ZNRD1 (NP_055411, 24 a.a. ~ 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GSVLPLPGAQDTVTCIRCGFNINVRDFEGKVVKTSVVFHQLGTAMPMSVEEGPECQGPVVDRRCPRCGHEGMAYHTRQMRSADEGQTVFYTCTNCKFQEKEDS | |
| 0.1 mg | |
| DNA replication Transcription Translation and Splicing | |
| 30834 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 6C12 | |
| Western Blot 1:500, ELISA | |
| NP_055411 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction