missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ ZN Carbol Fuchsin Stain

Product Code. 12916317
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
Unit Size:
1 L
2 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12916317 500 mL 500mL
12926317 1 L 1 L
12938637 2 L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12916317 Supplier Pro-Lab Diagnostics™ Supplier No. PL.7018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use as a primary stain for identifying acid-fast mycobacterium. Pro-Lab Diagnostics™ ZN Carbol Fuchsin Stain is a ready-to-use stain that is manufactured to the highest standards, according to all current regulatory requirements, and CE labelled.

Pro-Lab Diagnostics™ Microbiological Stain solutions are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all solutions offer excellent performance characteristics.

Acid-fast organisms, such as Mycobacterium, have wax-like, nearly impermeable cell walls that make them highly resistant to typical microbiological stains. They require staining via either the Ziehl-Neelsen (ZN) method, which requires heating, or the Kinyoun method, which does not. The basic method seen in Gram staining is used: a primary stain is applied, a differentiator removes the stain from non-acid-fast organisms and a counterstain is then applied to identify the decolored organisms.

  • Ready-to-use stain in convenient and economical sizes
  • The most common primary stain for use with the Ziehl-Neelsen method
  • Lipid-soluble red stain contains phenol, which helps the stain penetrate the cell wall 

Specifications

Type Carbol Fuchsin
Quantity 500 mL
Certifications/Compliance CE marked (98/78/EC)
Format Liquid
For Use With (Application) Staining of acid-fast mycobacterium using the Ziehl-Neelsen (ZN) method
Color Yellow/Magenta
Composition Ethanol 10 - <25% , Phenol 5 - <10% , Methanol 1 - <2.5% , Basic fuchsin 1 - <2.5%
Storage Requirements Keep at temperature not exceeding 20°C
Stain Type Acid Fast
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • ZN Carbol Fuchsin
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
  • P202-Do not handle until all safety precautions have been read and understood.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, phenol, methanol, basic fuchsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.