missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYND8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11633-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
| ZMYND8 | |
| Polyclonal | |
| Unconjugated | |
| ZMYND8 | |
| CTCL associated antigen se14 3, KIAA1125, PRKCBP1, PRO2893, RACK7, ZMYND8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 228880, 23613, 296374 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9ULU4 | |
| Zmynd8 | |
| ZMYND8 Fusion Protein Ag2207 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction