missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZMYND17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598471
This item is not returnable.
View return policy
Description
ZMYND17 Polyclonal Antibody for Western Blot, ELISA
ZMYND17 contains 1 MYND-type zinc finger. The exact function of ZMYND17 remains unknown.
Specifications
| ZMYND17 | |
| Polyclonal | |
| Unconjugated | |
| MSS51 | |
| Mss51; MSS51 mitochondrial translational activator; MSS51 mitochondrial translational activator homolog; putative protein MSS51 homolog, mitochondrial; Zinc finger MYND domain-containing protein 17; zinc finger, MYND domain containing 17; zinc finger, MYND-type containing 17; ZMYND17 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 118490 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.72 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q4VC12 | |
| MSS51 | |
| Recombinant Human Putative protein MSS51 homolog, mitochondrial protein (258-365AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction