missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZMYND11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112924
This item is not returnable.
View return policy
Description
ZMYND11 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
The protein encoded by this gene was first identified by its ability to bind the adenovirus E1A protein. The protein localizes to the nucleus. It functions as a transcriptional repressor, and expression of E1A inhibits this repression. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| ZMYND11 | |
| Polyclonal | |
| Unconjugated | |
| ZMYND11 | |
| 2210402G22Rik; adenovirus 5 E1A-binding protein; Bone morphogenetic protein receptor-associated molecule 1; BRAM1; BS69; MGC111056; MRD30; Protein BS69; RP11-486H9.1; Unknown (protein for MGC:137432); zinc finger MYND domain-containing protein 11; Zinc finger MYND domain-containing protein 11-like protein; zinc finger MYND-type containing 11; zinc finger, MYND domain containing 11; zinc finger, MYND-type containing 11; ZMYND11 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10771, 66505 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q15326, Q8R5C8 | |
| ZMYND11 | |
| Recombinant Human Zinc finger MYND domain-containing protein 11 protein (363-513AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction