missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYND11 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94400-0.1ml
This item is not returnable.
View return policy
Description
ZMYND11 Polyclonal antibody specifically detects ZMYND11 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ZMYND11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| adenovirus 5 E1A binding protein, Adenovirus 5 E1A-binding protein, bone morphogenetic protein receptor-associated molecule 1, BRAM1, MGC111056, MYND domain containing 11, Protein BS69, zinc finger MYND domain-containing protein 11, zinc finger, MYND-type containing 11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 278-567 of human ZMYND11 (NP_997644.2). PNHELVWAKMKGFGFWPAKVMQKEDNQVDVRFFGHHHQRAWIPSENIQDITVNIHRLHVKRSMGWKKACDELELHQRFLREGRFWKSKNEDRGEEEAESSISSTSNEQLKVTQEPRAKKGRRNQSVEPKKEEPEPETEAVSSSQEIPTMPQPIEKVSVSTQTKKLSASSPRMLHRSTQTTNDGVCQSMCHDKYTKIFNDFKDRMKSDHKRETERVVREALEKLRSEMEEEKRQAVNKAVANMQGEMDRKCKQVKEKCKEEFVEEIKKLATQHKQLISQTKKKQWVNTSLF | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Immunology | |
| 10771 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction