missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZMYM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599744
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZNF262.
Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. It has been found to be overexpressed in human lung adenocarcinomas and squamous cell carcinomas, and the overexpression of a fragment of the 3'UTR of the ZMYM4 mRNA termed Cell Death Inhibiting RNA (CDIR) protects HeLa cells from IFN-gamma-induced apoptosis, suggesting that ZMYM4 may play a role in tumorigenesis.
Specifications
| ZMYM4 | |
| Polyclonal | |
| Unconjugated | |
| ZMYM4 | |
| 6330503C17Rik; AI480785; AW493829; CDIR; cell death inhibiting RNA; D630001M21; Kiaa0425; mKIAA0425; MYM; MYM type 4; Zfp262; zinc finger MYM-type containing 4; zinc finger MYM-type protein 4; Zinc finger protein 262; zinc finger, MYM-type 4; Zmym4; ZNF198L3; ZNF262 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 313598, 67785, 9202 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2A791, Q5VZL5 | |
| ZMYM4 | |
| A synthesized peptide derived from human ZMYM4(Accession Q5VZL5), corresponding to amino acid residues S804-C854. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction