missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25742-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is located on the X chromosome and is subject to X inactivation. It is highly conserved in vertebrates and most abundantly expressed in the brain. The encoded protein is a component of histone deacetylase-containing multiprotein complexes that function through modifying chromatin structure to keep genes silent. A chromosomal translocation (X;13) involving this gene is associated with X-linked cognitive disability. Several alternatively spliced transcript variants have been found for this gene.Specifications
| ZMYM3 | |
| Polyclonal | |
| Unconjugated | |
| ZMYM3 | |
| DXS6673E, KIAA0385, MYM, XFIM, Zinc finger MYM type protein 3, Zinc finger protein 261, zinc finger, MYM type 3, ZMYM3, ZNF198L2, ZNF261 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56364, 9203 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14202, Q9JLM4 | |
| Zmym3 | |
| ZMYM3 Fusion Protein Ag22701 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction