missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM3 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76525PCP
This item is not returnable.
View return policy
Description
ZMYM3 Polyclonal antibody specifically detects ZMYM3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| ZMYM3 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the center of human ZMYM3. The immunogen is located within amino acids 220 - 270 of ZMYM3. | |
| 0.1 mL | |
| DNA Repair | |
| 9203 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction