missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM3 Antibody (PCRP-ZMYM3-2F10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08875MFV500
This item is not returnable.
View return policy
Description
ZMYM3 Monoclonal antibody specifically detects ZMYM3 in Human samples. It is validated for Flow Cytometry
Specifications
| ZMYM3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-ZMYM3-2F10 | |
| Flow Cytometry | |
| DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3 | |
| Recombinant full-length human ZMYM3 protein (Uniprot: Q14202) | |
| 0.1 mL | |
| DNA Repair | |
| 9203 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction