missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZMYM3 Polyclonal antibody specifically detects ZMYM3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | ZMYM3 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 18 amino acid synthetic peptide near the center of human ZMYM3. The immunogen is located within amino acids 220 - 270 of ZMYM3. |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?