missing translation for 'onlineSavingsMsg'
Learn More

ZMYM3 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-458CL1

Product Code. 30140767

  • £771.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZMYM3 Polyclonal antibody specifically detects ZMYM3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZMYM3
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CoraFluor 1
DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3
The immunogen this antibody was made to, maps to a region between residues 250-350 of human Zinc Finger Protein 261 using the NP_005087.1 entry Q8IY18 (GeneID 9203).
0.1 mL
DNA Repair
9203
Store at 4°C in the dark. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.