missing translation for 'onlineSavingsMsg'
Learn More

ZMYM1 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18432547 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432547 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432547 Supplier Novus Biologicals Supplier No. NBP176523R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZMYM1 Polyclonal antibody specifically detects ZMYM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYM1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias FLJ23151, zinc finger, MYM domain containing 1, zinc finger, MYM-type 1
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZMYM1 . The immunogen is located within amino acids 990 - 1040 of ZMYM1. Amino Acid Squence: WNEPLNETTAKHVQEFY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79830
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.