missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZMPSTE24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA512160
This item is not returnable.
View return policy
Description
ZMPSTE24 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy.
Specifications
| ZMPSTE24 | |
| Polyclonal | |
| Unconjugated | |
| ZMPSTE24 | |
| A530043O15Rik; CAAX prenyl protease 1 homolog; D030046F19; Face1; Face-1; Farnesylated proteins-converting enzyme 1; farnesylated-proteins converting enzyme 1; fi45a02; FLJ14968; HGPS; MADB; Prenyl protein-specific endoprotease 1; PRO1; STE24; Ste24p; wu:fi45a02; zgc:55655; zinc metallopeptidase STE24; zinc metallopeptidase STE24 homolog; zinc metallopeptidase, STE24; zinc metallopeptidase, STE24 homolog; zinc metalloproteinase (STE24 homolog, yeast); zinc metalloproteinase Ste24 homolog; zinc metalloproteinase, STE24 homolog; Zmpste24 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 10269 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O75844 | |
| ZMPSTE24 | |
| KLH conjugated synthetic peptide between 400-430 amino acids from the central region of human ZMPSTE24. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction