missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZMIZ2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13445787
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13445787 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13445787 Supplier Invitrogen™ Supplier No. PA531271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32. Predicted reactivity: Mouse (92%), Rat (93%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZIMP7, also known as ZMIZ2, is a novel PIAS (protein inhibitor of activated signal transducer and activator of transcription)-like protein and a transcriptional coactivator. ZIMP7 is expressed most abundantly in testis. The C-terminal proline-rich domain possesses a significant intrinsic transcriptional activity and this activity is inhibited by the N-terminus in the full-length ZIMP7. ZIMP7 and the related protein ZIMP10 interact with PIAS3 and enhances Androgen Receptor (AR)- mediated transcription. The interaction between ZIMP7 and SWI/SNF complex suggests a possible role for ZIMP7 in chromatin modification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMIZ2
Applications Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZMIZ2
Gene Accession No. Q8NF64
Gene Alias D11Bwg0280e; HRIHFB2007; hZIMP7; KIAA1886; NET27; PIAS-like protein Zimp7; TRAFIP20; Zimp7; zinc finger MIZ domain-containing protein 2; zinc finger MIZ-type containing 2; zinc finger, MIZ-type containing 2; Zmiz2
Gene Symbols ZMIZ2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZMIZ2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.