missing translation for 'onlineSavingsMsg'
Learn More

ZMIZ2 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18400296 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400296 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400296 Supplier Novus Biologicals Supplier No. NBP176552UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZMIZ2 Polyclonal antibody specifically detects ZMIZ2 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMIZ2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias hZIMP7, KIAA1886ZIMP7DKFZp761I2123, NET27, PIAS-like protein Zimp7, TRAFIP20, zinc finger MIZ domain-containing protein 2, zinc finger, MIZ-type containing 2
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human ZIMP7. The immunogen is located within amino acids 270 - 320 of ZIMP7. Amino Acid Squence: TAQFAPSPGQPPAPSPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83637
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.