missing translation for 'onlineSavingsMsg'
Learn More

ZMIZ1/Zimp10 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18432335 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432335 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432335 Supplier Novus Biologicals Supplier No. NBP176553AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZMIZ1/Zimp10 Polyclonal antibody specifically detects ZMIZ1/Zimp10 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMIZ1/Zimp10
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias FLJ13541, hZIMP10, KIAA1224Zimp10, MIZ, PIAS-like protein Zimp10, RAI17TRAFIP10, retinoic acid induced 17, Retinoic acid-induced protein 17, RP11-519K18.1, ZIMP10FLJ00092 protein, zinc finger MIZ domain-containing protein 1, zinc finger, MIZ-type containing 1, zinc finger-containing, Miz1, PIAS-like protein on chromosome 10
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human ZIMP10. The immunogen is located within amino acids 90 - 140 of ZIMP10. Amino Acid Squence: LRHQKSRQSDPPGKL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 57178
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.