missing translation for 'onlineSavingsMsg'
Learn More

ZMAT4 Antibody (OTI9E11), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£195.00 - £417.00

Specifications

Antigen ZMAT4
Clone OTI9E11
Dilution Western Blot 1:500-2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306214
View Documents
Novus Biologicals
NBP3-45007-30ul
30 μL
£195.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306732
View Documents
Novus Biologicals
NBP3-45007-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZMAT4 Monoclonal antibody specifically detects ZMAT4 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications

Specifications

ZMAT4
Western Blot 1:500-2000
Monoclonal
Purified
RUO
PBS (pH 7.3) 1% BSA, 50% glycerol
79698
IgG1
>95%
Antibody
OTI9E11
Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
FLJ13842, zinc finger matrin-type protein 4, zinc finger, matrin type 4, zinc finger, matrin-type 4
Full length human recombinant protein of human ZMAT4 (NP_078921) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.