missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMAT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10504-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein containing three zinc finger domains and a nuclear localization signal. The mRNA and the protein of this gene are upregulated by wildtype p53 and overexpression of this gene inhibits tumor cell growth, suggesting that this gene may have a role in the p53-dependent growth regulatory pathway. Alternative splicing of this gene results in two transcript variants encoding two isoforms differing in only one amino acid.Specifications
| ZMAT3 | |
| Polyclonal | |
| Unconjugated | |
| ZMAT3 | |
| p53 activated gene 608 protein, PAG608, WIG 1, WIG1, Zinc finger protein WIG 1, zinc finger, matrin type 3, ZMAT3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22401, 64393, 64394 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08781, O54836, Q9HA38 | |
| ZMAT3 | |
| ZMAT3 Fusion Protein Ag0780 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction