missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZMAT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103358
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZMAT3.
This gene encodes a protein containing three zinc finger domains and a nuclear localization signal. The mRNA and the protein of this gene are upregulated by wildtype p53 and overexpression of this gene inhibits tumor cell growth, suggesting that this gene may have a role in the p53-dependent growth regulatory pathway. Alternative splicing of this gene results in two transcript variants encoding two isoforms differing in only one amino acid.
Specifications
| ZMAT3 | |
| Polyclonal | |
| Unconjugated | |
| ZMAT3 | |
| FLJ12296; matrin type 3; MGC10613; p53 target zinc finger protein; p53-activated gene 608 protein; Pag608; UniProtKB:Q9HA38}; Wig1; WIG-1; WIG-1/PAG608 protein; wild-type p53-induced gene 1; wild-type p53-induced gene 1 protein; zinc finger matrin type 3; zinc finger matrin-type 3; zinc finger matrin-type protein 3; zinc finger protein WIG1; Zinc finger protein WIG-1; zinc finger, matrin type 3; zmat3; zmat3 {ECO:0000250 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22401, 64393 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54836, Q9HA38 | |
| ZMAT3 | |
| A synthesized peptide derived from human ZMAT3(Accession Q9HA38), corresponding to amino acid residues S133-N183. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction