missing translation for 'onlineSavingsMsg'
Learn More

ZM241385, MedChemExpress

Product Code. 16497626
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497626 100 mg 100mg
16483747 50 mg 50mg
16450258 5 mg 5mg
16417798 10 mg 10mg
16401296 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497626 Supplier Medchem Express Supplier No. HY19532
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ZM241385 is a potent, high affinity and selective adenosine A2a receptor (A2AR) antagonist with a Ki value of 1.4 nM.

ZM241385 is a potent, high affinity and selective adenosine A2a receptor (A2AR) antagonist with a Ki value of 1.4 nM.

Specifications

Chemical Name or Material ZM241385
CAS 139180-30-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H15N7O2
Quantity 100 mg
Solubility Information DMSO : ≥ 30 mg/mL (88.93 mM)
SMILES OC1=CC=C(CCNC2=NC3=NC(C4=CC=CO4)=NN3C(N)=N2)C=C1
Molecular Weight (g/mol) 337.34
Formula Weight 337.34
Percent Purity 98.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.