missing translation for 'onlineSavingsMsg'
Learn More

ZM 306416, MedChemExpress

Product Code. 16482007
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482007 50 mg 50mg
16475858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16482007 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY13785

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZM-306416 (CB 676475) is a potent inhibitor of VEGFR with IC50s of 0.1 and 2 μM for KDR and Flt, respectively. ZM-306416 is also a EGFR inhibitor with an IC50 of <10 nM.

ZM-306416 (CB 676475) is a potent inhibitor of VEGFR with IC50s of 0.1 and 2 μM for KDR and Flt, respectively. ZM-306416 is also a EGFR inhibitor with an IC50 of <10 nM.

Specifications

Chemical Name or Material ZM 306416
CAS 690206-97-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H13ClFN3O2
Quantity 50 mg
Synonym CB 676475
Solubility Information DMSO : 50 mg/mL (149.82 mM; Need ultrasonic)
SMILES COC1=CC2=NC=NC(NC3=C(F)C=C(Cl)C=C3)=C2C=C1OC
Molecular Weight (g/mol) 333.74
Formula Weight 333.74
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.