missing translation for 'onlineSavingsMsg'
Learn More

ZLN005, MedChemExpress

Product Code. 16437388
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16437388

Brand: Medchem Express HY17538

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZLN005 is a potent activator of peroxisome proliferator-activated receptor-γ coactivator-1α (PGC-1α).

ZLN005 is a potent activator of peroxisome proliferator-activated receptor-γ coactivator-1α (PGC-1α).

Specifications

Chemical Name or Material ZLN005
CAS 49671-76-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H18N2
Quantity 10 mg
Solubility Information DMSO : 22 mg/mL (87.88 mM; Need ultrasonic)
SMILES CC(C1=CC=C(C2=NC3=CC=CC=C3N2)C=C1)(C)C
Molecular Weight (g/mol) 250.34
Formula Weight 250.34
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.